missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2-(Boc-amino)cyclohexanecarboxylic acid, 97%
CAS: 209128-50-7 | C12H21NO4 | 243.303 g/mol
Supplier: Thermo Scientific Chemicals H6200706
| Quantity | 5 g |
|---|
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 209128-50-7 | |
| 243.303 | |
| QJEQJDJFJWWURK-RKDXNWHRSA-N | |
| 1512488 | |
| CC(C)(C)OC(=O)NC1CCCCC1C(=O)O |
| C12H21NO4 | |
| MFCD02094138 | |
| boc-1,2-trans-achc-oh, trans-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2r-boc-2-aminocyclohexane carboxylic acid, boc-trans-2-aminocyclohexanecarboxylic acid, trans-2-boc-amino cyclohexanecarboxylic acid, 1r,2r-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, trans-1-t-butyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, pubchem24192, trans-2-boc-amino-cyclohexanecarboxylic acid | |
| (1R,2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid |
Specifications
| trans-2-(Boc-amino)cyclohexanecarboxylic acid | |
| C12H21NO4 | |
| boc-1,2-trans-achc-oh, trans-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2r-boc-2-aminocyclohexane carboxylic acid, boc-trans-2-aminocyclohexanecarboxylic acid, trans-2-boc-amino cyclohexanecarboxylic acid, 1r,2r-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, trans-1-t-butyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, pubchem24192, trans-2-boc-amino-cyclohexanecarboxylic acid | |
| CC(C)(C)OC(=O)NC1CCCCC1C(=O)O | |
| 5 g | |
| 1512488 | |
| 97% |
| 209128-50-7 | |
| MFCD02094138 | |
| QJEQJDJFJWWURK-RKDXNWHRSA-N | |
| (1R,2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid | |
| 243.303 | |
| 243.29 | |
| Solid |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold; Store under Argon