missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Boc-amino)cyclohexanecarboxylic acid, 97%

CAS: 209128-50-7 | C12H21NO4 | 243.303 g/mol

Supplier:  Thermo Scientific Chemicals H6200703

Quantity 1 g

 View more versions of this product

Catalog No. AAH6200703

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

209128-50-7
243.303
QJEQJDJFJWWURK-RKDXNWHRSA-N
1512488
CC(C)(C)OC(=O)NC1CCCCC1C(=O)O
C12H21NO4
MFCD02094138
boc-1,2-trans-achc-oh, trans-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2r-boc-2-aminocyclohexane carboxylic acid, boc-trans-2-aminocyclohexanecarboxylic acid, trans-2-boc-amino cyclohexanecarboxylic acid, 1r,2r-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, trans-1-t-butyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, pubchem24192, trans-2-boc-amino-cyclohexanecarboxylic acid
(1R,2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid

Specifications

trans-2-(Boc-amino)cyclohexanecarboxylic acid
C12H21NO4
boc-1,2-trans-achc-oh, trans-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2r-boc-2-aminocyclohexane carboxylic acid, boc-trans-2-aminocyclohexanecarboxylic acid, trans-2-boc-amino cyclohexanecarboxylic acid, 1r,2r-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, trans-1-t-butyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, pubchem24192, trans-2-boc-amino-cyclohexanecarboxylic acid
CC(C)(C)OC(=O)NC1CCCCC1C(=O)O
1 g
1512488
97%
209128-50-7
MFCD02094138
QJEQJDJFJWWURK-RKDXNWHRSA-N
(1R,2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
243.303
243.29
Solid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions