missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Boc-amino)cyclohexanecarboxylic acid, 97%

Catalog No. p-7054100
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62007MD 250 mg
AAH6200703 1 g
AAH6200706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH62007MD Supplier Thermo Scientific Chemicals Supplier No. H62007MD

CAS: 209128-50-7 | C12H21NO4 | 243.303 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 209128-50-7
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD02094138
InChI Key QJEQJDJFJWWURK-RKDXNWHRSA-N
Synonym boc-1,2-trans-achc-oh, trans-2-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 1r,2r-boc-2-aminocyclohexane carboxylic acid, boc-trans-2-aminocyclohexanecarboxylic acid, trans-2-boc-amino cyclohexanecarboxylic acid, 1r,2r-2-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, trans-1-t-butyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, pubchem24192, trans-2-boc-amino-cyclohexanecarboxylic acid
PubChem CID 1512488
IUPAC Name (1R,2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCCC1C(=O)O

Specifications

Quantity 250 mg
Formula Weight 243.29
Percent Purity 97%
Physical Form Solid
Chemical Name or Material trans-2-(Boc-amino)cyclohexanecarboxylic acid
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only