missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH6263403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6263403 1 g
AAH6263406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6263403 Supplier Thermo Scientific Chemicals Supplier No. H6263403
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61935-48-6
Molecular Formula C15H19NO4
Molecular Weight (g/mol) 277.32
MDL Number MFCD03844616
InChI Key RPJMLWMATNCSIS-STQMWFEESA-N
Synonym z-1,2-trans-achc-oh, trans-1-benzyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, 1s,2s-2-benzyloxy carbonyl amino cyclohexane-1-carboxylic acid
PubChem CID 51341834
IUPAC Name (1S,2S)-2-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
SMILES C1CCC(C(C1)C(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

Quantity 1 g
Formula Weight 277.31
Percent Purity 97%
Physical Form Solid
Chemical Name or Material trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only