missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2,6-Difluorocinnamic Acid 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 122°C |
| Quantity | 5 g |
| Formula Weight | 184.14 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | trans-2,6-Difluorocinnamic Acid |
Safety and Handling
| TSCA | No |