missing translation for 'onlineSavingsMsg'
Learn More

trans-2,6-Difluorocinnamic Acid 95.0+%, TCI America™

Catalog No. D41151G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D41151G 1 g
D41155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D41151G Supplier TCI America Supplier No. D41151G
Only null left

Chemical Identifiers

CAS 102082-89-3
Molecular Formula C9H6F2O2
Molecular Weight (g/mol) 184.142
MDL Number MFCD00010319
InChI Key JMUOYANNVIFGFN-SNAWJCMRSA-N
Synonym 2,6-difluorocinnamic acid, trans-2,6-difluorocinnamic acid, e-3-2,6-difluorophenyl acrylic acid, 3-2,6-difluorophenyl acrylic acid, 2e-3-2,6-difluorophenyl prop-2-enoic acid, 2e-3-2,6-difluorophenyl acrylic acid, 3-2,6-difluoro-phenyl-acrylic acid, e-3-2,6-difluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-2,6-difluorophenyl-, 2e, rarechem bk hw 0116
PubChem CID 5372267
IUPAC Name (E)-3-(2,6-difluorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C(=C1)F)C=CC(=O)O)F

Specifications

Melting Point 122°C
Quantity 1 g
Formula Weight 184.14
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,6-Difluorocinnamic Acid