missing translation for 'onlineSavingsMsg'
Learn More

trans-2,5-Dichlorocinnamic Acid 96.0+%, TCI America™

$71.23 - $211.77

Chemical Identifiers

CAS 20595-47-5
Molecular Formula C9H6Cl2O2
Molecular Weight (g/mol) 217.045
MDL Number MFCD03002786
InChI Key LYYBVUOVYNSRSE-DAFODLJHSA-N
Synonym trans-3-(2,5-Dichlorophenyl)-2-propenoic Acid
PubChem CID 7014934
IUPAC Name (E)-3-(2,5-dichlorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1Cl)C=CC(=O)O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D37945G
View Documents
TCI America
D37945G
5 g
Each for $71.23
Only null left
 
D379425G
View Documents
TCI America
D379425G
25 g
Each for $211.77
Only null left
 
Description

Description

Specifications

Chemical Identifiers

20595-47-5
217.045
LYYBVUOVYNSRSE-DAFODLJHSA-N
7014934
C1=CC(=C(C=C1Cl)C=CC(=O)O)Cl
C9H6Cl2O2
MFCD03002786
trans-3-(2,5-Dichlorophenyl)-2-propenoic Acid
(E)-3-(2,5-dichlorophenyl)prop-2-enoic acid

Specifications

20595-47-5
White-Yellow
MFCD03002786
trans-3-(2,5-Dichlorophenyl)-2-propenoic Acid
C1=CC(=C(C=C1Cl)C=CC(=O)O)Cl
217.045
217.05
Crystalline Powder
192°C
C9H6Cl2O2
5 g
LYYBVUOVYNSRSE-DAFODLJHSA-N
(E)-3-(2,5-dichlorophenyl)prop-2-enoic acid
7014934
≥96.0% (GC,T)
trans-2,5-Dichlorocinnamic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'