missing translation for 'onlineSavingsMsg'
Learn More

trans-2,5-Dichlorocinnamic Acid 96.0+%, TCI America™

Catalog No. D37945G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37945G 5 g
D379425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37945G Supplier TCI America Supplier No. D37945G
Only null left

Chemical Identifiers

CAS 20595-47-5
Molecular Formula C9H6Cl2O2
Molecular Weight (g/mol) 217.045
MDL Number MFCD03002786
InChI Key LYYBVUOVYNSRSE-DAFODLJHSA-N
Synonym trans-3-(2,5-Dichlorophenyl)-2-propenoic Acid
PubChem CID 7014934
IUPAC Name (E)-3-(2,5-dichlorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1Cl)C=CC(=O)O)Cl

Specifications

Melting Point 192°C
Color White-Yellow
Quantity 5 g
Formula Weight 217.05
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,5-Dichlorocinnamic Acid