missing translation for 'onlineSavingsMsg'
Learn More

trans-2,3-Dibromo-2-butene-1,4-diol 99.0+%, TCI America™

Catalog No. D216925G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D216925G 25 g
D2169500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D216925G Supplier TCI America Supplier No. D216925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 21285-46-1
Molecular Formula C4H6Br2O2
Molecular Weight (g/mol) 245.90
MDL Number MFCD00004698
InChI Key MELXIJRBKWTTJH-ONEGZZNKSA-N
Synonym trans-2,3-dibromo-2-butene-1,4-diol, 2,3-dibromo-2-butene-1,4-diol, e-2,3-dibromobut-2-ene-1,4-diol, 2e-2,3-dibromobut-2-ene-1,4-diol, e-2,3-dibromo-2-butene-1,4-diol, 2,3-dibromobutene-1,4-diol, ccris 4781, 2-butene-1,4-diol, 2,3-dibromo-, 2e, 2-butene-1,4-diol, 2,3-dibromo, trans-2,3-dibromo-2-buten-1,4-diol
PubChem CID 641240
IUPAC Name (2E)-2,3-dibromobut-2-ene-1,4-diol
SMILES OC\C(Br)=C(/Br)CO

Specifications

Melting Point 114°C
Color White-Yellow
Quantity 25 g
Formula Weight 245.90
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-2,3-Dibromo-2-butene-1,4-diol
EINECSNumber (2)-3941
RTECSNumber EM6910000
TSCA Yes