missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-2,2-Dimethyl-3-heptene 99.0+%, TCI America™
Supplier: TCI America D12605ML
Specifications
| trans-2,2-Dimethyl-3-heptene | |
| Colorless | |
| MFCD00039961 | |
| 3295 | |
| CCCC=CC(C)(C)C | |
| 126.243 | |
| 126.24 | |
| Liquid |
| 19550-75-5 | |
| C9H18 | |
| 5 mL | |
| BQOCYCICSYUPRF-BQYQJAHWSA-N | |
| (E)-2,2-dimethylhept-3-ene | |
| 5352650 | |
| ≥99.0% (GC) |
Chemical Identifiers
| 19550-75-5 | |
| 126.243 | |
| BQOCYCICSYUPRF-BQYQJAHWSA-N | |
| (E)-2,2-dimethylhept-3-ene |
| C9H18 | |
| MFCD00039961 | |
| 5352650 | |
| CCCC=CC(C)(C)C |
Safety and Handling
EINECSNumber : (2)-0025
TSCA : No