missing translation for 'onlineSavingsMsg'
Learn More

trans-1-Octenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

$111.04 - $111.04

Chemical Identifiers

CAS 83947-55-1
Molecular Formula C14H27BO2
Molecular Weight (g/mol) 238.178
MDL Number MFCD03453667
InChI Key KQTOSGTXAFJZSJ-VAWYXSNFSA-N
Synonym trans-4,4,5,5-tetramethyl-2-oct-1-enyl-1,3,2-dioxaborolane, 1-octenylboronic acid pinacol ester, 1-octen-1-ylboronic acid, pinacol ester, trans-1-octenylboronic acid pinacol ester, trans-4,4,5,5-tetramethyl-2-1-octenyl-1,3,2-dioxaborolane, e-1-octenylboronic acid pinacol ester, trans-1-octen-1-ylboronic acid pinacol ester, trans-1-oct-1-en-1-yl boronic acid, pinacol ester, trans-4,4,5,5-tetramethyl-2-oct-1-en-1-yl-1,3,2-dioxaborolane
PubChem CID 5706555
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-oct-1-enyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=CCCCCCC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1969703
View Documents
Thermo Scientific Chemicals
L1969703
1 g
Each for $111.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

83947-55-1
238.178
KQTOSGTXAFJZSJ-VAWYXSNFSA-N
5706555
B1(OC(C(O1)(C)C)(C)C)C=CCCCCCC
C14H27BO2
MFCD03453667
trans-4,4,5,5-tetramethyl-2-oct-1-enyl-1,3,2-dioxaborolane, 1-octenylboronic acid pinacol ester, 1-octen-1-ylboronic acid, pinacol ester, trans-1-octenylboronic acid pinacol ester, trans-4,4,5,5-tetramethyl-2-1-octenyl-1,3,2-dioxaborolane, e-1-octenylboronic acid pinacol ester, trans-1-octen-1-ylboronic acid pinacol ester, trans-1-oct-1-en-1-yl boronic acid, pinacol ester, trans-4,4,5,5-tetramethyl-2-oct-1-en-1-yl-1,3,2-dioxaborolane
4,4,5,5-tetramethyl-2-[(E)-oct-1-enyl]-1,3,2-dioxaborolane

Specifications

83947-55-1
1.446
1 g
trans-4,4,5,5-tetramethyl-2-oct-1-enyl-1,3,2-dioxaborolane, 1-octenylboronic acid pinacol ester, 1-octen-1-ylboronic acid, pinacol ester, trans-1-octenylboronic acid pinacol ester, trans-4,4,5,5-tetramethyl-2-1-octenyl-1,3,2-dioxaborolane, e-1-octenylboronic acid pinacol ester, trans-1-octen-1-ylboronic acid pinacol ester, trans-1-oct-1-en-1-yl boronic acid, pinacol ester, trans-4,4,5,5-tetramethyl-2-oct-1-en-1-yl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C=CCCCCCC
238.178
238.18
trans-1-Octenylboronic acid pinacol ester
C14H27BO2
MFCD03453667
7291389
KQTOSGTXAFJZSJ-VAWYXSNFSA-N
4,4,5,5-tetramethyl-2-[(E)-oct-1-enyl]-1,3,2-dioxaborolane
5706555
97%
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only