missing translation for 'onlineSavingsMsg'
Learn More

trans-1-Octenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog No. AAL1969703
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1969703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1969703 Supplier Thermo Scientific Chemicals Supplier No. L1969703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 83947-55-1 | Molecular Formula: C14H27BO2 | Molecular Weight: 238.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 83947-55-1
Molecular Formula C14H27BO2
Molecular Weight (g/mol) 238.178
MDL Number MFCD03453667
InChI Key KQTOSGTXAFJZSJ-VAWYXSNFSA-N
Synonym trans-4,4,5,5-tetramethyl-2-oct-1-enyl-1,3,2-dioxaborolane, 1-octenylboronic acid pinacol ester, 1-octen-1-ylboronic acid, pinacol ester, trans-1-octenylboronic acid pinacol ester, trans-4,4,5,5-tetramethyl-2-1-octenyl-1,3,2-dioxaborolane, e-1-octenylboronic acid pinacol ester, trans-1-octen-1-ylboronic acid pinacol ester, trans-1-oct-1-en-1-yl boronic acid, pinacol ester, trans-4,4,5,5-tetramethyl-2-oct-1-en-1-yl-1,3,2-dioxaborolane
PubChem CID 5706555
IUPAC Name 4,4,5,5-tetramethyl-2-[(E)-oct-1-enyl]-1,3,2-dioxaborolane
SMILES B1(OC(C(O1)(C)C)(C)C)C=CCCCCCC

Specifications

Refractive Index 1.446
Quantity 1 g
Beilstein 7291389
Formula Weight 238.18
Percent Purity 97%
Chemical Name or Material trans-1-Octenylboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only