missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,4-Dichlorocyclohexane 99.0+%, TCI America™
Supplier: TCI America D14255G
Specifications
| trans-1,4-Dichlorocyclohexane | |
| 103°C | |
| 60°C | |
| MFCD00070474 | |
| WQTINZDWAXJLGH-UHFFFAOYSA-N | |
| 1,4-dichlorocyclohexane | |
| 519304 | |
| ≥99.0% (GC) |
| 16890-91-8 | |
| White | |
| C6H10Cl2 | |
| 5 g | |
| ClC1CCC(Cl)CC1 | |
| 153.05 | |
| 153.05 | |
| Crystalline Powder |
Chemical Identifiers
| 16890-91-8 | |
| 153.05 | |
| WQTINZDWAXJLGH-UHFFFAOYSA-N | |
| 1,4-dichlorocyclohexane |
| C6H10Cl2 | |
| MFCD00070474 | |
| 519304 | |
| ClC1CCC(Cl)CC1 |
Safety and Handling
TSCA : No