missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,4-Cyclohexanedicarboxylic Acid 97.0+%, TCI America™
Supplier: TCI America C0475100G
Specifications
| trans-1,4-Cyclohexanedicarboxylic Acid | |
| White | |
| MFCD00066203 | |
| 1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane | |
| C1CC(CCC1C(=O)O)C(=O)O | |
| 172.18 | |
| 172.18 | |
| Crystalline Powder |
| 619-82-9 | |
| C8H12O4 | |
| 100 g | |
| PXGZQGDTEZPERC-UHFFFAOYSA-N | |
| cyclohexane-1,4-dicarboxylic acid | |
| 14106 | |
| ≥97.0% (GC,T) |
Chemical Identifiers
| 619-82-9 | |
| 172.18 | |
| PXGZQGDTEZPERC-UHFFFAOYSA-N | |
| 14106 | |
| C1CC(CCC1C(=O)O)C(=O)O |
| C8H12O4 | |
| MFCD00066203 | |
| 1,4-cyclohexanedicarboxylic acid, trans-1,4-cyclohexanedicarboxylic acid, cis-1,4-cyclohexanedicarboxylic acid, cis-cyclohexane-1,4-dicarboxylic acid, trans-cyclohexane-1,4-dicarboxylic acid, hexahydroterephthalic acid, trans-hexahydroterephthalic acid, trans-1,4-cyclohexanedicarboxybic acid, cis-hexahydroterephthalic acid, 1,4-dicarboxycyclohexane | |
| cyclohexane-1,4-dicarboxylic acid |
Safety and Handling
EINECSNumber : (3)-2423&(3)-2446
TSCA : No