missing translation for 'onlineSavingsMsg'
Learn More

trans-1,4-Bis(aminomethyl)cyclohexane 98.0+%, TCI America™

Catalog No. B51475G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51475G 5 g
B51471G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51475G Supplier TCI America Supplier No. B51475G

Chemical Identifiers

CAS 10029-07-9
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.25
MDL Number MFCD00039463,MFCD20037427,MFCD20617651
InChI Key OXIKYYJDTWKERT-UHFFFAOYSA-N
Synonym trans-1,4-Di(aminomethyl)cyclohexane, trans-Hexahydro-p-xylylenediamine
PubChem CID 17354
IUPAC Name 1-[4-(aminomethyl)cyclohexyl]methanamine
SMILES NCC1CCC(CN)CC1

Specifications

Color Colorless
Boiling Point 118°C
Quantity 5 g
UN Number 2922
Formula Weight 142.25
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material trans-1,4-Bis(aminomethyl)cyclohexane
EINECSNumber (3)-4352
TSCA No