missing translation for 'onlineSavingsMsg'
Learn More

trans-1,3-Diphenyl-2-propen-1-ol 97.0+%, TCI America™

Catalog No. p-7127512
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28085G 5 g
D280825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28085G Supplier TCI America Supplier No. D28085G

Chemical Identifiers

CAS 62668-02-4
Molecular Formula C15H14O
Molecular Weight (g/mol) 210.276
MDL Number MFCD00239426
InChI Key ORACYDGVNJGDMI-VAWYXSNFSA-N
PubChem CID 5712032
IUPAC Name (E)-1,3-diphenylprop-2-en-1-ol
SMILES C1=CC=C(C=C1)C=CC(C2=CC=CC=C2)O

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 5 g
Formula Weight 210.28
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,3-Diphenyl-2-propen-1-ol
TSCA No
Recommended Storage Refrigerator