missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dimethylcyclohexane 99.0+%, TCI America™

Catalog No. D069725ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D069725ML 25 mL
D06975ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D069725ML Supplier TCI America Supplier No. D069725ML
Only null left

Chemical Identifiers

CAS 6876-23-9
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00001505
InChI Key KVZJLSYJROEPSQ-YUMQZZPRSA-N
Synonym trans-Hexahydro-o-xylene
PubChem CID 252360
IUPAC Name (1S,2S)-1,2-dimethylcyclohexane
SMILES CC1CCCCC1C

Specifications

Color Colorless
Boiling Point 124°C
Quantity 25 mL
UN Number 2263
Formula Weight 112.22
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-1,2-Dimethylcyclohexane
EINECSNumber (3)-2227
TSCA Yes