missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Dimethylcyclohexane 99.0+%, TCI America™
$150.98 - $395.17
Chemical Identifiers
| CAS | 6876-23-9 |
|---|---|
| Molecular Formula | C8H16 |
| Molecular Weight (g/mol) | 112.216 |
| MDL Number | MFCD00001505 |
| InChI Key | KVZJLSYJROEPSQ-YUMQZZPRSA-N |
| Synonym | trans-Hexahydro-o-xylene |
| PubChem CID | 252360 |
| IUPAC Name | (1S,2S)-1,2-dimethylcyclohexane |
| SMILES | CC1CCCCC1C |
Chemical Identifiers
| 6876-23-9 | |
| 112.216 | |
| KVZJLSYJROEPSQ-YUMQZZPRSA-N | |
| 252360 | |
| CC1CCCCC1C |
| C8H16 | |
| MFCD00001505 | |
| trans-Hexahydro-o-xylene | |
| (1S,2S)-1,2-dimethylcyclohexane |
Specifications
| 6876-23-9 | |
| 124°C | |
| MFCD00001505 | |
| 2263 | |
| KVZJLSYJROEPSQ-YUMQZZPRSA-N | |
| (1S,2S)-1,2-dimethylcyclohexane | |
| 252360 | |
| ≥99.0% (GC) | |
| trans-1,2-Dimethylcyclohexane |
| Colorless | |
| C8H16 | |
| 5 mL | |
| trans-Hexahydro-o-xylene | |
| CC1CCCCC1C | |
| 112.216 | |
| 112.22 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-2227
TSCA : Yes