missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichloroethylene (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. D036825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D036825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D036825G Supplier TCI America Supplier No. D036825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 156-60-5
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.94
MDL Number MFCD00062942
InChI Key KFUSEUYYWQURPO-OWOJBTEDSA-N
Synonym trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
PubChem CID 638186
ChEBI CHEBI:29027
IUPAC Name (1E)-1,2-dichloroethene
SMILES Cl\C=C\Cl

Specifications

Color Colorless
Boiling Point 48°C
Quantity 25 g
UN Number 1150
Formula Weight 96.94
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material trans-1,2-Dichloroethylene (stabilized with MEHQ)
EINECSNumber (2)-0103
RTECSNumber KV9400000
TSCA Yes