missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichloroethylene, 99+%, stabilized

Catalog No. AC406840250
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC406840250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC406840250 Supplier Thermo Scientific Chemicals Supplier No. 406840250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 156-60-5 | C2H2Cl2 | 96.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 156-60-5
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.94
MDL Number MFCD00062942
InChI Key KFUSEUYYWQURPO-OWOJBTEDSA-N
Synonym trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
PubChem CID 638186
ChEBI CHEBI:29027
IUPAC Name (E)-1,2-dichloroethene
SMILES Cl\C=C\Cl

Specifications

Melting Point -50°C
Color Colorless
Density 1.2600g/mL
Boiling Point 48°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.4450 to 1.447
Linear Formula CHClCHCl
Quantity 25 g
Beilstein 01, 188
Specific Gravity 1.26
Merck Index 15, 87
Solubility Information Solubility in water: immiscible
Stabilizer 85 to 115ppm MEHQ
Formula Weight 96.94
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material trans-1, 2-Dichloroethylene, 99.7%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if inhaled.
Harmful to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away
EINECSNumber 205-860-2
RTECSNumber KV9400000
TSCA TSCA

RUO – Research Use Only