missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Dichloroethylene, 98%, stab. with 4-methoxyphenol

Catalog No. p-7046331
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1399414 25 g
AAL1399422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1399414 Supplier Thermo Scientific Chemicals Supplier No. L1399414
Only null left

CAS: 156-60-5 | C2H2Cl2 | 96.94 g/mol

trans-1,2-Dichloroethylene is used in the preparation of solvents and in chemical mixtures. It also acts as a synthetic agent and active constituent of perfumes. Further, it is used for electronics cleaning, precision cleaning and metal cleaning activities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Dichloroethylene is used in the preparation of solvents and in chemical mixtures. It also acts as a synthetic agent and active constituent of perfumes. Further, it is used for electronics cleaning, precision cleaning and metal cleaning activities.

Solubility
Miscible with ethanol, ethyl ether, acetone, benzene and chloroform. Immiscible with water.

Notes
Light, air and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents and bases.

Chemical Identifiers

CAS 156-60-5
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.94
MDL Number MFCD00062942
InChI Key KFUSEUYYWQURPO-OWOJBTEDSA-N
Synonym trans-1,2-dichloroethylene, trans-1,2-dichloroethene, e-1,2-dichloroethene, e-1,2-dichloroethylene, trans-dichloroethylene, trans-acetylene dichloride, 1,2-trans-dichloroethylene, acetylene dichloride, ethene, 1,2-dichloro-, 1e, dioform
PubChem CID 638186
ChEBI CHEBI:29027
IUPAC Name (E)-1,2-dichloroethene
SMILES Cl\C=C\Cl

Specifications

Melting Point -50°C
Density 1.252
Boiling Point 47°C to 49°C
Flash Point 2°C (35°F)
Refractive Index 1.4455
Quantity 25 g
UN Number UN1150
Beilstein 1420761
Merck Index 14,92
Solubility Information Miscible with ethanol,ethyl ether,acetone,benzene and chloroform. Immiscible with water.
Formula Weight 96.94
Percent Purity 98%
Chemical Name or Material trans-1,2-Dichloroethylene, stabilized with 4-methoxyphenol
Show More Show Less
Hazard Category H225-H302+H332
Hazard Statement GHS H Statement
H225-H302-H332
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P304+P340-P312-P330-P370+P378q-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1,2-DICHLOROETHYLENE
EINECSNumber 205-860-2
RTECSNumber KV9400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only