missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Di(2-thienyl)ethylene 98.0+%, TCI America™

Catalog No. p-7127095
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20741G 1 g
D20745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D20741G Supplier TCI America Supplier No. D20741G
Only null left

Chemical Identifiers

CAS 13640-78-3
Molecular Formula C10H8S2
Molecular Weight (g/mol) 192.294
MDL Number MFCD00145416
InChI Key AYBFWHPZXYPJFW-AATRIKPKSA-N
PubChem CID 5375272
IUPAC Name 2-[(E)-2-thiophen-2-ylethenyl]thiophene
SMILES C1=CSC(=C1)C=CC2=CC=CS2

Specifications

Melting Point 133°C
Color Yellow
Quantity 1 g
Formula Weight 192.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material trans-1,2-Di(2-thienyl)ethylene