missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Di(2-thienyl)ethylene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 133°C |
| Color | Yellow |
| Quantity | 5 g |
| Formula Weight | 192.29 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | trans-1,2-Di(2-thienyl)ethylene |
Safety and Handling
| TSCA | No |