missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Cyclohexanediol, 98%
CAS: 1460-57-7 | C6H12O2 | 116.16 g/mol
$139.10 - $420.14
Chemical Identifiers
| CAS | 1460-57-7 |
|---|---|
| Molecular Formula | C6H12O2 |
| Molecular Weight (g/mol) | 116.16 |
| MDL Number | MFCD00063611 |
| InChI Key | PFURGBBHAOXLIO-PHDIDXHHSA-N |
| Synonym | trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol |
| PubChem CID | 92886 |
| ChEBI | CHEBI:16931 |
| IUPAC Name | (1R,2R)-cyclohexane-1,2-diol |
| SMILES | O[C@@H]1CCCC[C@H]1O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0708814
|
Thermo Scientific Chemicals
L0708814 |
25 g |
Each for $139.10
|
|
|||||
|
AAL0708822
|
Thermo Scientific Chemicals
L0708822 |
100 g |
Each for $420.14
|
|
|||||
Description
trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationstrans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.
Solubility
Soluble in chloroform and methanol.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
| 1460-57-7 | |
| 116.16 | |
| PFURGBBHAOXLIO-PHDIDXHHSA-N | |
| 92886 | |
| (1R,2R)-cyclohexane-1,2-diol |
| C6H12O2 | |
| MFCD00063611 | |
| trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
| CHEBI:16931 | |
| O[C@@H]1CCCC[C@H]1O |
Specifications
| 1460-57-7 | |
| 231°C to 233°C | |
| C6H12O2 | |
| 25 g | |
| trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol | |
| PFURGBBHAOXLIO-PHDIDXHHSA-N | |
| (1R,2R)-cyclohexane-1,2-diol | |
| 92886 | |
| 116.16 | |
| trans-1,2-Cyclohexanediol |
| 101°C to 106°C | |
| 134°C (273°F) | |
| MFCD00063611 | |
| 3193810 | |
| Soluble in chloroform and methanol. | |
| O[C@@H]1CCCC[C@H]1O | |
| 116.16 | |
| CHEBI:16931 | |
| 98% |
Safety and Handling
EINECSNumber : 215-956-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only