missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Cyclohexanediol, 98%

Catalog No. AAL0708814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0708814 25 g
AAL0708822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0708814 Supplier Thermo Scientific Chemicals Supplier No. L0708814

CAS: 1460-57-7 | C6H12O2 | 116.16 g/mol

trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
trans-1,2-Cyclohexanediol is used as an organic chemical synthesis intermediate.

Solubility
Soluble in chloroform and methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 1460-57-7
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00063611
InChI Key PFURGBBHAOXLIO-PHDIDXHHSA-N
Synonym trans-1,2-cyclohexanediol, trans-cyclohexane-1,2-diol, 1r,2r-cyclohexane-1,2-diol, 1r,2r-trans-1,2-cyclohexanediol, 1,2-cyclohexanediol, 1r,2r-rel, trans-1,2-dihydroxycyclohexane, 1,2-cyclohexanediol, trans, l6tj aq bq&&trans, trans-1,2-cyclohexandiol, 1r,2r-1,2-cyclohexanediol
PubChem CID 92886
ChEBI CHEBI:16931
IUPAC Name (1R,2R)-cyclohexane-1,2-diol
SMILES O[C@@H]1CCCC[C@H]1O

Specifications

Melting Point 101°C to 106°C
Boiling Point 231°C to 233°C
Flash Point 134°C (273°F)
Quantity 25 g
Beilstein 3193810
Solubility Information Soluble in chloroform and methanol.
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material trans-1,2-Cyclohexanediol
EINECSNumber 215-956-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only