missing translation for 'onlineSavingsMsg'
Learn More

(-)-trans-1,2-Cyclohexanedicarboxylic Anhydride 98.0+%, TCI America™

Catalog No. p-7124968
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1418100MG 100 mg
C14181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1418100MG Supplier TCI America Supplier No. C1418100MG

Chemical Identifiers

CAS 31982-85-1
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
MDL Number MFCD00674195
InChI Key MUTGBJKUEZFXGO-WDSKDSINSA-N
PubChem CID 12261141
IUPAC Name (3aS,7aS)-3a,4,5,6,7,7a-hexahydro-2-benzofuran-1,3-dione
SMILES C1CCC2C(C1)C(=O)OC2=O

Specifications

Melting Point 166°C
Color White
Quantity 100 mg
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (-)-trans-1,2-Cyclohexanedicarboxylic Anhydride
EINECSNumber (3)-2416
TSCA No