missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-trans-1,2-Cyclohexanedicarboxylic Anhydride 98.0+%, TCI America™
$178.95 - $915.25
Chemical Identifiers
| CAS | 71749-03-6 |
|---|---|
| Molecular Formula | C8H10O3 |
| Molecular Weight (g/mol) | 154.17 |
| MDL Number | MFCD00064345,MFCD00674194 |
| InChI Key | MUTGBJKUEZFXGO-UHFFFAOYNA-N |
| PubChem CID | 10942665 |
| IUPAC Name | octahydro-2-benzofuran-1,3-dione |
| SMILES | O=C1OC(=O)C2CCCCC12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C1417100MG
|
TCI America
C1417100MG |
100 mg |
Each for $178.95
|
|
|||||
|
C14171G
|
TCI America
C14171G |
1 g |
Each for $915.25
|
|
|||||
Chemical Identifiers
| 71749-03-6 | |
| 154.17 | |
| MUTGBJKUEZFXGO-UHFFFAOYNA-N | |
| octahydro-2-benzofuran-1,3-dione |
| C8H10O3 | |
| MFCD00064345,MFCD00674194 | |
| 10942665 | |
| O=C1OC(=O)C2CCCCC12 |
Specifications
| 71749-03-6 | |
| White-Yellow | |
| MFCD00064345,MFCD00674194 | |
| MUTGBJKUEZFXGO-UHFFFAOYNA-N | |
| octahydro-2-benzofuran-1,3-dione | |
| 10942665 | |
| ≥98.0% (GC) | |
| (+)-trans-1,2-Cyclohexanedicarboxylic Anhydride |
| 165°C | |
| C8H10O3 | |
| 100 mg | |
| O=C1OC(=O)C2CCCCC12 | |
| 154.17 | |
| 154.17 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-2416
TSCA : No