missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+)-trans-1,2-Cyclohexanedicarboxylic Anhydride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 165°C |
| Color | White-Yellow |
| Quantity | 1 g |
| Formula Weight | 154.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (+)-trans-1,2-Cyclohexanedicarboxylic Anhydride |
Safety and Handling
| EINECSNumber | (3)-2416 |
| TSCA | No |