missing translation for 'onlineSavingsMsg'
Learn More

(+)-trans-1,2-Cyclohexanedicarboxylic Anhydride 98.0+%, TCI America™

Catalog No. C1417100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1417100MG 100 mg
C14171G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1417100MG Supplier TCI America Supplier No. C1417100MG
Only null left

Chemical Identifiers

CAS 71749-03-6
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00064345,MFCD00674194
InChI Key MUTGBJKUEZFXGO-UHFFFAOYNA-N
PubChem CID 10942665
IUPAC Name octahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2CCCCC12

Specifications

Melting Point 165°C
Color White-Yellow
Quantity 100 mg
Formula Weight 154.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (+)-trans-1,2-Cyclohexanedicarboxylic Anhydride
EINECSNumber (3)-2416
TSCA No