missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Cyclohexanedicarboxylic acid, 98%

Catalog No. AAL1128806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1128806 5 g
AAL1128814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1128806 Supplier Thermo Scientific Chemicals Supplier No. L1128806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2305-32-0 | C8H12O4 | 172.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2305-32-0
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
MDL Number MFCD00001464
InChI Key QSAWQNUELGIYBC-PHDIDXHHSA-N
Synonym 1r,2r-cyclohexane-1,2-dicarboxylic acid, trans-1,2-cyclohexanedicarboxylic acid, trans-cyclohexane-1,2-dicarboxylic acid, 1r,2r---1,2-cyclohexanedicarboxylic acid, 1r,2r-1,2-cyclohexanedicarboxylic acid, 1,2-cyclohexanedicarboxylic acid, 1r,2r-rel, 1,2-cyclohexanedicarboxylic acid, 1r,2r, 1r.2r-cyclohexane-1,2-dicarboxylicacid, 1,2-cyclohexanedicarboxylic acid, trans, pubchem19603
PubChem CID 720895
IUPAC Name (1R,2R)-cyclohexane-1,2-dicarboxylic acid
SMILES C1CCC(C(C1)C(=O)O)C(=O)O

Specifications

Melting Point 220°C to 221°C
Quantity 5 g
Beilstein 3200609
Formula Weight 172.18
Percent Purity 98%
Chemical Name or Material trans-1,2-Cyclohexanedicarboxylic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 218-975-8
RTECSNumber GU9065000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only