missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1,2-Cyclohexanediamine 97.0+%, TCI America™
$83.89 - $417.56
Chemical Identifiers
| CAS | 1121-22-8 |
|---|---|
| Molecular Formula | C6H14N2 |
| Molecular Weight (g/mol) | 114.192 |
| MDL Number | MFCD00063747 |
| InChI Key | SSJXIUAHEKJCMH-PHDIDXHHSA-N |
| Synonym | 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine |
| PubChem CID | 43806 |
| IUPAC Name | (1R,2R)-cyclohexane-1,2-diamine |
| SMILES | C1CCC(C(C1)N)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
C112025ML
|
TCI America
C112025ML |
25 mL |
Each for $83.89
|
|
|||||
|
C1120250ML
|
TCI America
C1120250ML |
250 mL |
Each for $417.56
|
|
|||||
Chemical Identifiers
| 1121-22-8 | |
| 114.192 | |
| SSJXIUAHEKJCMH-PHDIDXHHSA-N | |
| 43806 | |
| C1CCC(C(C1)N)N |
| C6H14N2 | |
| MFCD00063747 | |
| 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine | |
| (1R,2R)-cyclohexane-1,2-diamine |
Specifications
| 1121-22-8 | |
| 80°C | |
| MFCD00063747 | |
| 2735 | |
| SSJXIUAHEKJCMH-PHDIDXHHSA-N | |
| (1R,2R)-cyclohexane-1,2-diamine | |
| 43806 | |
| ≥97.0% (GC,T) | |
| trans-1,2-Cyclohexanediamine |
| Yellow | |
| C6H14N2 | |
| 25 mL | |
| 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine | |
| C1CCC(C(C1)N)N | |
| 114.192 | |
| 114.19 | |
| Liquid |
Safety and Handling
EINECSNumber : (3)-4230
TSCA : Yes