missing translation for 'onlineSavingsMsg'
Learn More

trans-1,2-Cyclohexanediamine 97.0+%, TCI America™

Catalog No. C112025ML
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C112025ML 25 mL
C1120250ML 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C112025ML Supplier TCI America Supplier No. C112025ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1121-22-8
Molecular Formula C6H14N2
Molecular Weight (g/mol) 114.192
MDL Number MFCD00063747
InChI Key SSJXIUAHEKJCMH-PHDIDXHHSA-N
Synonym 1r,2r---1,2-diaminocyclohexane, 1r,2r-cyclohexane-1,2-diamine, trans-1,2-diaminocyclohexane, trans-1,2-cyclohexanediamine, trans-cyclohexane-1,2-diamine, 1r,2r-1,2-diaminocyclohexane, 1r,2r---1,2-cyclohexanediamine, 1r,2r-diaminocyclohexane, r-dach, 1r,2r-1,2-cyclohexanediamine
PubChem CID 43806
IUPAC Name (1R,2R)-cyclohexane-1,2-diamine
SMILES C1CCC(C(C1)N)N

Specifications

Color Yellow
Boiling Point 80°C
Quantity 25 mL
UN Number 2735
Formula Weight 114.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material trans-1,2-Cyclohexanediamine
EINECSNumber (3)-4230
TSCA Yes