missing translation for 'onlineSavingsMsg'
Learn More

Tranexamic acid, 98+%

Catalog No. AC228042500
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228042500 250 g Glass Bottle
AC228040500 50 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC228042500 Supplier Thermo Scientific Chemicals Supplier No. 228042500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1197-18-8 | C8H15NO2 | 157.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1197-18-8
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00001466
InChI Key GYDJEQRTZSCIOI-UHFFFAOYSA-N
Synonym tranexamic acid, cyklokapron, tranexamsaeure, trans amcha, tranhexamic acid, trans-4-aminomethyl cyclohexanecarboxylic acid, transamin, amstat, tamcha, amikapron
PubChem CID 5526
IUPAC Name 4-(aminomethyl)cyclohexane-1-carboxylic acid
SMILES C1CC(CCC1CN)C(=O)O

Specifications

Melting Point >300°C
pH 6.5 to 8 (5% soln.)
Color White
Infrared Spectrum Authentic
Loss on Drying 1% max.
Assay Percent Range 98+%
Packaging Glass Bottle
Linear Formula H2NCH2C6H10CO2H
Merck Index 15, 9730
Solubility Information Solubility in water: 166g/L. Other solubilities: slightly soluble in alcohol and ether,practically insoluble in most organic solvents
Quantity 250 g
Formula Weight 157.21
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material Tranexamic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
EINECSNumber 214-818-2

RUO – Research Use Only