missing translation for 'onlineSavingsMsg'
Learn More

Tralkoxydim 95.0+%, TCI America™

Catalog No. T2916200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T2916200MG 200 mg
T29161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T2916200MG Supplier TCI America Supplier No. T2916200MG

Chemical Identifiers

CAS 87820-88-0
Molecular Formula C20H27NO3
Molecular Weight (g/mol) 329.44
MDL Number MFCD00145415
InChI Key SOTLWPHEAQOHHC-UHFFFAOYSA-N
PubChem CID 91746
IUPAC Name 2-[1-(ethoxyamino)propylidene]-5-(2,4,6-trimethylphenyl)cyclohexane-1,3-dione
SMILES CCC(=C1C(=O)CC(CC1=O)C2=C(C=C(C=C2C)C)C)NOCC

Specifications

Melting Point 107°C
Quantity 200 mg
UN Number 3077
Formula Weight 329.44
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Tralkoxydim
RTECSNumber GW7191600
TSCA No