missing translation for 'onlineSavingsMsg'
Learn More

Tragacanth powder

CAS: 9000-65-1

$134.00 - $386.60

Chemical Identifiers

CAS 9000-65-1
MDL Number MFCD00131255
Synonym Gum tragacanth
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1850222
View Documents
Thermo Scientific Chemicals
A1850222
100 g
Each for $134.00
Only null left
 
AAA1850236
View Documents
Thermo Scientific Chemicals
A1850236
500 g
Each for $386.60
Only null left
 
Description

Description

Tragacanth powder is used as an emulsifier, thickener, stabilizer in food and pharmaceutical industry. It acts as a binder used in the preparation of artist's pastels. It is used as a natural hair shampoo. It plays an important role as freezing-embedding medium for tissue preservation. It is used as a demulcent in cough and cold preparations as well as to manage diarrhea.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

9000-65-1
MFCD00131255
Gum tragacanth

Specifications

9000-65-1
14,4581
Slightly soluble in water.
Odorless
MFCD00131255
Gum tragacanth
100 g
Tragacanth powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-552-5

RTECSNumber : XW7750000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only