missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tragacanth, Powder, NF, Spectrum™ Chemical
Description
Spectrum™ Chemical Tragacanth, Powder, NF is used as an emulsifier, thickener, stabilizer, and texturant additive. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 1 |
| Packaging | Poly Bottle |
| Quantity | 500 g |
| Grade | NF |