missing translation for 'onlineSavingsMsg'
Learn More

Pyramex Safety Products GL102 Medium Duty Synthetic Leather Mechanics Gloves

Catalog No. 19555039
Change view
Click to view available options
Size:
Small
Medium
Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19555039 Small
19555038 Medium
19555037 Large
19555040 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19555039 Supplier Pyramex Safety Products Supplier No. GL102S
Only null left

Gloves feature thicker embossed neoprene on the knuckles for protection, with touchscreen compatibility on two fingers and thumb.

  • Double layer spandex with mesh for comfort and breathability
  • Silicone palm patch for anti-slip and enhanced grip capabilities
  • Thicker embossed neoprene on knuckles for additional protection
  • Touch screen function on 2 fingers and thumb

Specifications

Back Material Mesh
Coating Material Silicone
Color Black
Hand Specific
Material Synthetic Leather, Spandex
Product Type Mechanic's Gloves
Size Small
Finger Style Full
For Use With (Application) Mechanics, Construction, Fabrication, and Assembly
Glove Cuff Style Hook and Loop
Series GL102
Show More Show Less