missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ p-Terphenyl-d14 Solution, 2,000 μg/mL in dichloromethane, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 1718-51-0 |
| MDL Number | MFCD00084184 |
| Type | Food and Cosmetic Component Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 2,000 μg/mL in dichloromethane |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC