missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ MISA Group 17 Non-Halogen Organic Mix, Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Quantity | 1 mL |
| Packaging | Ampule |
| UN Number | UN1230 - Class 3 - PG 2 - Methanol, solution |
| Grade | Trace CERT |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC