missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ L-Cystine, MilliporeSigma™ Supelco™

Catalog No. 111014668
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014668 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014668 Supplier Merck Emd Millipore Supplier No. 49603100MG
Only null left

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
Product Line TraceCERT
CAS 56-89-3
Physical Form Neat
Linear Formula [-SCH2 CH(NH2)CO2 H]2
MDL Number MFCD00064228
Melting Point >240°C
Synonym (R,R)-3,3 ′-Dithiobis(2-aminopropionicacid)
Type Certified Reference Material
Quantity 100 mg
Formula Weight 240.30
Grade TraceCERT™ Analytical Standard
Show More Show Less

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC