missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ HDI-DBA-d9 Oligomers Standard, Certified Reference Material, in Methanol (varied conc.), MilliporeSigma™ Supelco™
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| Synonym | HDI adducts; Hexamethylene diisocyanate dibutylamine-d9 oligomers standard |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC