missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Fragrance Allergen Mix A2, Certified Reference Material, MilliporeSigma™ Supelco™
This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.
$703.38 - $1432.73
Specifications
| Flash Point | -33°C (Closed Cup) |
|---|---|
| UN Number | UN 2398 3/PGII |
| Grade | Trace CERT™ |
Description
- 40 Components in Tert-Butyl Methyl Ether
Specifications
| -33°C (Closed Cup) | |
| UN 2398 3/PGII | |
| Trace CERT™ |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC