missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Fragrance Allergen Mix A1, Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Flash Point | -33°C (Closed Cup) |
| Quantity | 2 mL |
| UN Number | UN 2398 3/PGII |
| Grade | Trace CERT |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC