missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ EPA VOC Mix 8, Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Organic Pollutant Standards |
| Flash Point | 11°C (Closed Cup, Not Applicable) |
| Packaging | Ampule |
| Quantity | 1 mL |
| UN Number | UN1230 - Class 3 - PG 2 - Methanol, solution |
| Synonym | Volatile Organic Compounds Mix 8 |
| Grade | Trace CERT |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC