missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ EPA 524 Internal Standard Mix, Certified Reference Material, MilliporeSigma™ Supelco™
Specifications
Specifications
| Chemical Name or Material | Organic Pollutant Standards |
| Flash Point | 9.7°C (Closed Cup) |
| Packaging | Ampule |
| Quantity | 1 mL |
| UN Number | UN1230 - Class 3 - PG 2 - Methanol, solution |
| Grade | Trace CERT |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC