missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ (±)-Diclofop, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022767
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022767 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022767 Supplier Merck Emd Millipore Supplier No. 7401150MG

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Specifications

CAS 40843-25-2
Flash Point Not Applicable
Molecular Formula C15H12Cl2O4
Linear Formula C15H12Cl2O4
Quantity 50 mg
MDL Number MFCD00152216
Synonym (RS)-2-[4-(2,4-Dichlorophenoxy)phenoxy]propionic acid
Formula Weight 327.16
Grade Trace CERT

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC