missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TraceCERT™ Decafluorotriphenylphosphine Solution, 25 mg/mL in dichloromethane, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| Packaging | Ampule |
| Product Line | TraceCERT |
| CAS | 5074-71-5 |
| Synonym | 2,3,4,5,6,2 ′,3 ′,4 ′,5 ′,6 ′-Decafluorotriphenylphosphine; Bis(pentafluorophenyl)phenylphosphine |
| Type | Organic Pollutant Standard |
| Quantity | 1 mL |
| Grade | TraceCERT™ Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 25 mg/mL in dichloromethane |
TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC