missing translation for 'onlineSavingsMsg'
Learn More

TraceCERT™ β-Cyfluthrin, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111022813
Change view
Click to view available options
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
111022813 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022813 Supplier Merck Emd Millipore Supplier No. 9322350MG
Only null left

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from a NMI. Certified content incl. uncertainty and expiry date are stated on the enclosed certificate.

Specifications

CAS 1820573-27-0
Flash Point Not Applicable
Molecular Formula C22H18Cl2FNO3
Linear Formula C22H18Cl2FNO3
Quantity 50 mg
MDL Number MFCD00078636
Formula Weight 434.29
Grade Trace CERT

TraceCERT is a registered trademark of Sigma-Aldrich Co. LLC