missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Platinum Ultra-Accurate Digital Thermometer

Catalog No. p-4581517
Change view
Click to view available options
Temperature Range (Metric):
-200°C to +500°C
Model:
Handle Probe
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric) Model
15081102 -200°C to +500°C Handle Probe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15081102
Only null left

Includes

Probe, cable, Plastic carrying case, traceable certificate and 9V battery

High-precision unit is designed for years of reliable service—even in the severest environments.

High-precision unit is designed for years of reliable service—even in the severest environments. Fisher Scientific™ Traceable™ Platinum Ultra-Accurate Digital Thermometers have platinum sensor which ensures linear response and high accuracy across the entire range. Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens and electronics manufacturing.

  • Platinum sensor insures linear response and high accuracy across the entire range
  • Ideal for foods, pharmaceuticals, petroleum products, wastewater, gas systems, soil, ovens, electronics manufacturing
  • Membrane-covered keys are splash-proof and easy to operate
  • HOLD allows user to freeze display for reading later
  • Displays data in °F or °C
  • Computer output supplied with unit
  • Handle Probe: (range –200 to 500°C): Fast-response, waterproof, 4-wire platinum/stainless-steel 316 probe, 0.16-inch diameter x 7-inch stem length, overall length 11.5 inches; probe tip penetration of 0.3-inch, 6-foot cable
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory

Specifications

Certifications/Compliance ISO 17025, A2LA, NIST
Depth (English) 0.5 in.
Depth (Metric) 1.27 cm
Diameter (English) Probe 0.16 in.
Diameter (Metric) Probe 0.4 cm
Dimensions (L x W x H) 11.5 x 3.5 x 5.5 in. (29.21 x 8.89 x 13.97 cm)
For Use With (Application) Measuring temperatures in gas systems, reagents, chemical solutions, soil, water baths, ovens, incubators, pharmaceuticals, foods and wastewater
Height (English) 5.5 in.
Height (Metric) 13.97 cm
Includes Stainless Steel probe with handle, cable, computer output capabilities (software package must be purchased separately), Plastic carrying case, traceable certificate and 9V battery
Length (English) Cable 6 ft.
Length (English) Probe 7 in.
Length (Metric) 292 mm
Length (Metric) Cable 182.88 cm
Length (Metric) Probe 17.7 cm
Material Platinum, Stainless Steel
Model Handle Probe
Resolution 0.001°C
Weight (English) 8 oz.
Weight (Metric) 226.7 g
Width (English) 3.5 in.
Product Type Digital Thermometer
Length (English) 11.5 in.
Max. Temperature (English) 932°F
Max. Temperature (Metric) 500°C
Min. Temperature (English) -328°F
Min. Temperature (Metric) -200°C
Temperature Range (Metric) -200°C to +500°C
Calibrated Yes
Accuracy ±0.05°C
Display Digital LCD
Width (Metric) 8.89 cm
Show More Show Less