Learn More
Research Products Corporation Traceable 4 Channel Timer
Catalog No. 50476593
Encompass
Its microcomputer chip permits setting 4 separate channels simultaneously in any combination of countdown or count-up (stopwatch). Set any countdown time from 99 hours, 59 minutes, 59 seconds to 1 second with an accuracy of 0.01%. When zero is reached, the timer starts counting up to show time elapsed since alarming. Additional features include stopwatch, time-out, and time-of-day clock. Finger-size keys make it easy to set times and change channels.
An individually-numbered Traceable Certificate provided with each unit, assures accuracy from our ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement.
Supplied with Spring fastener, flip-open stand, magnet, opening for lanyard, battery, and Traceable Certificate.
Catalog No. 50476593 Supplier Research Products Corporation Supplier No. RPI 825004
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Its microcomputer chip permits setting 4 separate channels simultaneously in any combination of countdown or count-up (stopwatch). Set any countdown time from 99 hours, 59 minutes, 59 seconds to 1 second with an accuracy of 0.01%. When zero is reached, the timer starts counting up to show time elapsed since alarming. Additional features include stopwatch, time-out, and time-of-day clock. Finger-size keys make it easy to set times and change channels.
An individually-numbered Traceable Certificate provided with each unit, assures accuracy from our ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement.
Supplied with Spring fastener, flip-open stand, magnet, opening for lanyard, battery, and Traceable Certificate.